Learn More
Perkin Elmer US LLC Replacement Filament Assembly for the T-9 Ion Source
Catalog No. 502113805
Encompass_Preferred
Calion standards are premixed and adsorbed into a solid phase. These standards are ideal for in field use because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration.
Catalog No. 50-211-3805 Supplier Perkin Elmer US LLC Supplier No. NTSS0070034
Calion standards are premixed and adsorbed into a solid phase. These standards are ideal for in field use because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration because they contain no liquids. Calion PV Mixes are used for Torion T-9 calibration, including both mass and GC retention time calibrations. The combination of Custodion SPME sampling with Calion standards provides robust and rapid capability for in-field calibration.