Learn More
Perkin Elmer US LLC Replacement Filters for Helium Cell Gas Filtration System
Supplier:  Perkin Elmer US LLC N8146005
Encompass_Preferred
Note: These filters are not for use with oxygen. Compatible with ICP-MS instruments such as the NexION 1000/2000/350/300.
Catalog No. 50-211-7296