Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Replacement Fingerprint Ink Rubber Rollers
SDP

Catalog No. 22039048 Shop All Mopec Products
Change view
Click to view available options
Width (English):
2 in.
3 in.
4 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
22-039-048 4 in.
22-039-046 2 in.
22-039-047 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-039-048 Supplier Mopec Supplier No. FS821
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fits Roll-Ease and Steel-Grip ink roller models.

  • Dimension
    • Diameter: 1.1875 in.
  • Options and Accessories
    • Roll-ease fingerprint ink rollers (FS816-FS818)
    • Steel-grip fingerprint ink rollers (FS822 and FS823)
  • Material
    • Rubber

Specifications

Product Type Replacement Rubber Roller
Width (English) 4 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.