Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dentec Safety™ Replacement Fit Test Sensitivity Ampules
SDP

Catalog No. 19222547 Shop All Dentec Safety Specialists Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Masks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-222-547 Masks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-222-547 Supplier Dentec Safety™ Supplier No. 79262
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Replacement fit test sensitivity ampules, 2.5 cc.

  • Sensitivity replacement fit test ampules
  • Volume of 2.5 cubic centimetrs
  • Clear/translucent
  • Meets the NIOSH standard

Specifications

Product Type Respirator Accessory
For Use With (Equipment) Masks
Model 79262
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.