Learn More
Research Products International Corp Replacement Foot Plate Kit for Vortex-Genie 2
SDP
Catalog No. 50550207
Encompass
This kit contains the Bottom Closure with Feet and (4) Screws. The Bottom Closure can be easily installed using either a 1/4" nut driver or pliers. Will fit onto Vortex-Genie 1, Vortex-Genie Pulse, Disruptor Genie, and Vortex-Genie 2 family of mixers.
Catalog No. 50-550-207 Supplier Research Products International Corp Supplier No. 155325
This kit contains the Bottom Closure with Feet and (4) Screws. The Bottom Closure can be easily installed using either a 1/4" nut driver or pliers. Will fit onto Vortex-Genie 1, Vortex-Genie Pulse, Disruptor Genie, and Vortex-Genie 2 family of mixers.