Learn More
Chemglass Life Sciences Replacement Glass Shaft, 100mL and 250mL Internal Spinner Flask
SDP
Catalog No. 501532192
Encompass_Preferred
Replacement components for CLS-1414 and CLS-1415 Spinner Flasks.The Complete Impeller Assembly consists of top cap, silicone liner, glass rod, compression fitting magnet holder, paddle and magnetic stir bar.
Catalog No. 50-153-2192 Supplier Chemglass Life Sciences Supplier No. CLS142620
Replacement components for CLS-1414 and CLS-1415 Spinner Flasks.The Complete Impeller Assembly consists of top cap, silicone liner, glass rod, compression fitting magnet holder, paddle and magnetic stir bar.