Learn More
Chemglass Life Sciences Replacement Glass Shaft, 7mm, 25mL Standard Spinner Flask
SDP
Catalog No. 501532172
Encompass
Replacement Components for CLS-1410- and CLS-1411-Series Spinner Flasks.The Replacement Shaft consists of the glass shaft only.
Catalog No. 50-153-2172 Supplier Chemglass Life Sciences Supplier No. CLS142520
Replacement Components for CLS-1410- and CLS-1411-Series Spinner Flasks.The Replacement Shaft consists of the glass shaft only.