Learn More
World Precision Instrument Replacement Heating Filament (large gauge) for DMF1000 Microforg
SDP
Catalog No. 502521196
Encompass
Can be reformed into a U shape for fabrication of pipettes, air forming of patch pipettes, and other applications.
Catalog No. 50-252-1196 Supplier World Precision Instrument Supplier No. DMF1000H5
Can be reformed into a U shape for fabrication of pipettes, air forming of patch pipettes, and other applications.