Learn More
TA Instruments Replacement ICTAC Certified Curie Point Reference Material, 153°C, Magnetic, for Temperature Calibration of TGA Instruments
Catalog No. 5000580616 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 153°C is a magnetic reference material used for temperature calibration of TGA instruments.
Catalog No. 50-005-80616 Supplier TA Instruments Supplier No. 952500.901
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 153°C is a magnetic reference material used for temperature calibration of TGA instruments.