Learn More
TA Instruments Replacement ICTAC Certified Curie Point Reference Material 554°C, Magnetic Reference Material, Temperature Calibration, TGA Instruments, Replacement ICTAC Certified Curie Point Reference Material 554°C
Catalog No. 5000580888 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 554°C is a magnetic reference material for temperature calibration of TGA instruments. It allows users to confirm the accuracy of TGA instrument temperature readings reliably.
Catalog No. 50-005-80888 Supplier TA Instruments Supplier No. 952508.901
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 554°C is a magnetic reference material for temperature calibration of TGA instruments. It allows users to confirm the accuracy of TGA instrument temperature readings reliably.