Learn More
TA Instruments Replacement ICTAC Certified Curie Point Reference Material 746°C, Magnetic Reference Material, Temperature Calibration, TGA Instruments, Replacement ICTAC Certified Curie Point Reference Material 746°C
Catalog No. 5000580614 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 746°C is a magnetic reference material for temperature calibration of TGA instruments. It supports consistent and precise calibration for high-temperature thermogravimetric analysis.
Catalog No. 50-005-80614 Supplier TA Instruments Supplier No. 952512.901
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 746°C is a magnetic reference material for temperature calibration of TGA instruments. It supports consistent and precise calibration for high-temperature thermogravimetric analysis.