Learn More
TA Instruments Replacement ICTAC Certified Curie Point Reference Material 931°C, Magnetic Reference Material, Temperature Calibration, TGA Instruments, Replacement ICTAC Certified Curie Point Reference Material 931°C
Catalog No. 5000580686 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 931°C is a magnetic reference material for temperature calibration of TGA instruments. It ensures accurate temperature calibration for experiments requiring high thermal precision.
Catalog No. 50-005-80686 Supplier TA Instruments Supplier No. 952516.901
Waters TA Instruments Replacement ICTAC Certified Curie Point Reference Material 931°C is a magnetic reference material for temperature calibration of TGA instruments. It ensures accurate temperature calibration for experiments requiring high thermal precision.