Learn More
World Precision Instrument Replacement Input Kit for PicoPumps
SDP
Catalog No. 502521470
Encompass
Accessories for the popular microinjection PicoPumps
This kit includes 10' of hard tubing, and NPT fitting and a vacuum fitting, which may be used with the WPI PicoPumps that are so popular for microinjection.
Catalog No. 50-252-1470 Supplier World Precision Instrument Supplier No. 3316
Accessories for the popular microinjection PicoPumps
This kit includes 10' of hard tubing, and NPT fitting and a vacuum fitting, which may be used with the WPI PicoPumps that are so popular for microinjection.