Learn More
Electron Microscopy Sciences Replacement Mesh Inserts, Set of 8
SDP
Catalog No. 501898860
Encompass
Set of 8 phosphor-bronzed screen inserts for micro-sieve in the following mesh: 25, 35, 45, 60, 80, 120, 170, and 320.
Catalog No. 50-189-8860 Supplier Electron Microscopy Sciences Supplier No. 5011100
Set of 8 phosphor-bronzed screen inserts for micro-sieve in the following mesh: 25, 35, 45, 60, 80, 120, 170, and 320.