Learn More
Chemglass Life Sciences Replacement Stainless-Steel Retention Kit for Chemglass High-Vacuum Stopcocks, 17/40 Plug Size (CG-469-21)
SDP
Catalog No. 502733526
Encompass_Preferred
Replacement S.S. retention kit (CG-469-21) contains a stainless-steel spring and clip designed to restore the constant-pressure retention device on compatible high-vacuum glass stopcocks. Supplied individually.
  • Restores constant tension on the glass plug.
  • Includes one stainless-steel spring.
  • Includes one stainless-steel clip.
  • For 17/40 plug size.
  • Sold individually.
Catalog No. 50-273-3526 Supplier Chemglass Life Sciences Supplier No. CG46921
Replacement S.S. retention kit (CG-469-21) contains a stainless-steel spring and clip designed to restore the constant-pressure retention device on compatible high-vacuum glass stopcocks. Supplied individually.
  • Restores constant tension on the glass plug.
  • Includes one stainless-steel spring.
  • Includes one stainless-steel clip.
  • For 17/40 plug size.
  • Sold individually.