Learn More
American International Electric REPLACEMENT KT FOR AIE-400
Supplier:  American International Electric K400P
Encompass_Preferred
This item has a minimum qty of 3 per supplier requirements.
American International Electric K400H 2 Element Wires, 2 Upper Non-Stick Fluoropolymer, 1 Lower Non-Stick Fluoropolymer Tape (2mm) for 16" Hand Sealer. This heat sealing repair kit is for AIE-400H machines.
Catalog No. NC1299441