Learn More
World Precision Instrument Replacement Halogen Lamp for Z-LITE
SDP
Catalog No. NC9955882
Encompass
  • 150W, 21V
  • 3350K, ultra bright
  • 40 hour average life
  • For use with the NOVA, Z-LITE and MSI-150
Catalog No. NC9955882 Supplier World Precision Instrument Supplier No. EJA
  • 150W, 21V
  • 3350K, ultra bright
  • 40 hour average life
  • For use with the NOVA, Z-LITE and MSI-150