Learn More
Molecular Devices Molecular Devices Laptop, Dell Latitude D620, 1.83 GHz, 2 GB RAM
Catalog No. 5000014382
Encompass_Preferred
The Molecular Devices PC Laptop Bundle (0310-4874) provides a ready-to-use laptop configured for instrument control, data acquisition, and basic office tasks when used with microplate readers.
  • Dual-core 1.83 GHz processor.
  • 2 GB RAM.
  • 80 GB hard drive.
  • 24x CD-RW/DVD optical drive.
  • 14.1 in WXGA display.
  • Includes office productivity and PDF software.
  • Configured for microplate reader instrument control and data acquisition.
Catalog No. 50-000-14382 Supplier Molecular Devices Supplier No. 03104874
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices PC Laptop Bundle (0310-4874) provides a ready-to-use laptop configured for instrument control, data acquisition, and basic office tasks when used with microplate readers.
  • Dual-core 1.83 GHz processor.
  • 2 GB RAM.
  • 80 GB hard drive.
  • 24x CD-RW/DVD optical drive.
  • 14.1 in WXGA display.
  • Includes office productivity and PDF software.
  • Configured for microplate reader instrument control and data acquisition.