Learn More
Fisherbrand™ Replacement Lamp for Shimadzu Spectrophotometer
Catalog No. 14500726 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Replacement Detector Lamp
Catalog No. Product Type
14-500-726 Replacement Detector Lamp
1 options
Catalog No. 14-500-726
Fisherbrand™ Replacement Lamp for Shimadzu Spectrophotometer has 2000 hours or 12 months shelf life.
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-726 is comparable to Shimadzu part 062-65055-05, 65063-01.
Compliance
WEEE and ROHS compliant
Specifications
Light Source Deuterium
For Use With (Equipment) Shimadzu Spectrophotometer
Product Type Replacement Detector Lamp