Learn More
Perkin Elmer US LLC Laser Head (HeNe) and Laser Power Supply Replacement Laser Kit for Spectrum One System
Catalog No. 509043202
Encompass_Preferred
These kits are required for all the PerkinElmer FT-IR Spectrometers to keep the optical path free from moisture. It is recommended that the laser head is replaced every six months. Follow the instructions in the appropriate instrument manual to install the laser head.
Catalog No. 50-904-3202 Supplier Perkin Elmer US LLC Supplier No. L1200253
These kits are required for all the PerkinElmer FT-IR Spectrometers to keep the optical path free from moisture. It is recommended that the laser head is replaced every six months. Follow the instructions in the appropriate instrument manual to install the laser head.