Learn More
Thermo Scientific™ Replacement Lid Assemblies and Seals for Fiberlite™ F14-6 x 250y Fixed Angle Rotors
Catalog No. 022062034 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Fiberlite F14S-6x250y
Catalog No. For Use With (Equipment)
02-206-2034 Fiberlite F14S-6x250y
1 options
Catalog No. 02-206-2034 Supplier Thermo Scientific™ Supplier No. 022062034
Add to Cart
Add to Cart
Use these replacement lid assemblies and seals with Thermo Scientific™ Fiberlite™ F14-6 x 250y Fixed Angle Rotors.
Use these Thermo Scientific™ Replacement Lid Assemblies and Seals for Fiberlite™ F14-6 x 250y with the accompanying fixed angle rotors.
Includes:
O-Ring Refresher Kits include five O-rings, three O-ring grease packets and 4 oz. FiberClene Lab Cleaner (017-0024)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Fiberlite F14S-6x250y
Description For FiberLite F14S-6x250y
Product Type Lid O-ring