Learn More
Perkin Elmer US LLC Replacement Lower Die for Sealer
Catalog No. 502111987
Encompass_Preferred
The Volatile Sample Pan Sealer is used to seal volatile, viscous, non-corrosive samples amd can be used in either Thermal Analysis or Elemental Analysis investigations.
Catalog No. 50-211-1987 Supplier Perkin Elmer US LLC Supplier No. 02191295
The Volatile Sample Pan Sealer is used to seal volatile, viscous, non-corrosive samples amd can be used in either Thermal Analysis or Elemental Analysis investigations.