Learn More
DWK Life Sciences Replacement metal GL 45 cap for Stainless Steel Laboratory Bottle
SDP
Catalog No. 02900138 Shop All DWK Life Sciences Products
Change view
Click to view available options
Thread Size (Metric):
99 mm
Catalog No. Thread Size (Metric)
02-900-138 99 mm
1 options
Catalog No. 02-900-138 Supplier DWK Life Sciences Supplier No. 299112808
Add to Cart
Add to Cart
Corrosion and damage resistant stainless steel laboratory bottle.
The unbreakable steel bottle is ideal for storage applications where glass is not applicable, due to the risk of breakage or chemical incompatibility. Manufactured from corrosion resistant AISI Type 316L (1.4404) Stainless steel, and hygienically constructed with all welded seams. The bottle has a smooth inner surface finish that corresponds to IIIc (DIN 17441) with rounded inner edges for ease of cleaning. Polished and brushed durable exterior finish. The bottle has a GL 45 thread with a built-in pouring lip. Supplied without a cap, Stainless steel cap is available. It is fully compatible with all the GL 45 caps.
  • Manufactured from corrosion resistant AISI Type 316L (1.4404) Stainless steel, and hygienically constructed with all welded seams
  • Polished and brushed durable exterior finish
  • Due to the smooth inner surface that corresponds to IIIc (DIN 17441) with rounded inner edges the bottle can be easily cleaned
  • GL 45 thread with a built-in pouring lip
  • Fully compatible with all the GL 45 caps
  • Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Storage, transport and safe handling of hazardous, dangerous and valuable materials; Storage and transport of light-sensitive materials
Thread Size (Metric) 99 mm
Neck Finish 99 mm