Learn More
Synthware™ Replacement Metering Stopcock
GREENER_CHOICESDP
Catalog No. 31502009 Shop All Kemtech Products
Change view
Click to view available options
Plug Bore:
2 mm
4 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Bore
31-502-009 2 mm
31-502-010 4 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-502-009 Supplier Synthware™ Supplier No. S191125
Add to Cart
Add to Cart
Stopcock comes with a threaded PTFE needle valve for the fine control of flow through the bore of the stopcock plug.
Specifications
Type Metering Stopcock
Plug Bore 2 mm
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
For Use With (Equipment) Metering Valve
Plug Material PTFE