Learn More
Perkin Elmer US LLC Replacement Needle Assembly for PinAAcle Plastic Neb, Organics
Catalog No. 502104365
Encompass_Preferred
Replacement needle assembly for Plastic High-Sensitivity PinAAcle AA nebulizers. This needle assembly has a Platinum/Iridium needle and Kalrez O-rings making it compatible with Aqueous or Organic solutions.
Catalog No. 50-210-4365 Supplier Perkin Elmer US LLC Supplier No. N3160154
Replacement needle assembly for Plastic High-Sensitivity PinAAcle AA nebulizers. This needle assembly has a Platinum/Iridium needle and Kalrez O-rings making it compatible with Aqueous or Organic solutions.