Learn More
TA Instruments Replacement O-Ring for Double Gap Concentric Cylinder Cup, Compatible with HA Aluminum P/N 546050.901 or Stainless Steel P/N 546045.901, Rated up to 200 Degrees Celsius with Peltier or Electrically Heated Jackets
Catalog No. 5000580862 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement O-Ring For Double Gap Concentric Cylinder Cup is for use with double gap concentric cylinder cups (HA Aluminum, P/N 546050.901 or Stainless Steel, P/N 546045.901). It is rated for use up to 200 degrees Celsius when used with HR X0, DHR, and AR-Series Concentric Cylinder Peltier Jackets (P/N 533201.901) or Electrically Heated Jackets (P/N 547310.901).
Catalog No. 50-005-80862 Supplier TA Instruments Supplier No. 578096.001
Waters TA Instruments Replacement O-Ring For Double Gap Concentric Cylinder Cup is for use with double gap concentric cylinder cups (HA Aluminum, P/N 546050.901 or Stainless Steel, P/N 546045.901). It is rated for use up to 200 degrees Celsius when used with HR X0, DHR, and AR-Series Concentric Cylinder Peltier Jackets (P/N 533201.901) or Electrically Heated Jackets (P/N 547310.901).