Learn More
TA Instruments Replacement O-Ring for Standard and Grooved Concentric Cylinder Cups with Cap, Compatible with HA Aluminum P/N 545700.901 or Stainless Steel P/N 545696.901, Grooved Cup P/N 545610.901, Rated up to 200 Degrees Celsius
Catalog No. 5000580704 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Replacement O-Ring For Standard And Grooved Concentric Cylinder Cups With Cap is for use with standard concentric cylinder cups (HA Aluminum, P/N 545700.901 or Stainless Steel, P/N 545696.901) and grooved concentric cylinder cups (P/N 545610.901). Rated for use up to 200 degrees Celsius when used with HR X0, DHR, and AR-Series Concentric Cylinder Peltier Jackets (P/N 533201.901) or Electrically Heated Jackets (P/N 547310.901).
Catalog No. 50-005-80704 Supplier TA Instruments Supplier No. 578148.001
Waters TA Instruments Replacement O-Ring For Standard And Grooved Concentric Cylinder Cups With Cap is for use with standard concentric cylinder cups (HA Aluminum, P/N 545700.901 or Stainless Steel, P/N 545696.901) and grooved concentric cylinder cups (P/N 545610.901). Rated for use up to 200 degrees Celsius when used with HR X0, DHR, and AR-Series Concentric Cylinder Peltier Jackets (P/N 533201.901) or Electrically Heated Jackets (P/N 547310.901).