Learn More
Perkin Elmer US LLC Replacement OilExpress Flowcell - ZnSe Windows
Supplier:  Perkin Elmer US LLC L1250220
Encompass_Preferred
This replacement cell is compatible with the Frontier OilExpress and Spectrum 100.
Catalog No. 50-211-3586