Learn More
Electron Microscopy Sciences Replacement Oval Graver
SDP
Catalog No. 50996718
Encompass
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.
Catalog No. 50-996-718 Supplier Electron Microscopy Sciences Supplier No. 6209715
Hexagonal anodized aluminum handles with hardened tool steel tips that can be sharpend or pointed. OAL is 140 mm.