Learn More
Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
Catalog No. 01910741 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Coulometric Reagent Set
Drying Tube
Septa
Solvent Drain/Delivery Tube
Titration Vessel
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-910-741 Drying Tube
01-910-738 Coulometric Cell without Diaphragm
01-910-737 Coulometric Cell with Diaphragm
01-911-144 Coulometric Reagent Set
01-910-735 Titration Vessel
01-910-744 Septa
01-910-747 Solvent Drain/Delivery Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-910-741 Supplier Mettler Toledo™ Supplier No. 51108733
Add to Cart
Edge
Add to Cart
Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator
Coulometric Cells
  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents
TRUSTED_SUSTAINABILITY
Specifications
Type Drying Tube
Material Glass