Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
GSA_VA

Catalog No. 01910744 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Coulometric Reagent Set
Drying Tube
Septa
Solvent Drain/Delivery Tube
Titration Vessel
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
01-910-744 Septa
01-910-738 Coulometric Cell without Diaphragm
01-910-737 Coulometric Cell with Diaphragm
01-911-144 Coulometric Reagent Set
01-910-735 Titration Vessel
01-910-747 Solvent Drain/Delivery Tube
01-910-741 Drying Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-910-744 Supplier Mettler Toledo™ Supplier No. 51108740
Add to Cart
Add to Cart

Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator

Coulometric Cells

  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Septa
For Use With (Equipment) C20 Coulometric Titrator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.