Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mettler Toledo™ Replacement Parts for C20 Karl Fischer™ Autotitrator
GSA_VA

Catalog No. p-4684528 Shop All METTLER TOLEDO Products
Click to view available options
Type:
Coulometric Cell with Diaphragm
Coulometric Cell without Diaphragm
Coulometric Reagent Set
Drying Tube
Septa
Solvent Drain/Delivery Tube
Titration Vessel

Accessories For use with Mettler Toledo C20 Karl Fischer™ Autotitrator

Coulometric Cells

  • Coulometer cells with or without diaphragm
  • Cell without diaphragm is almost maintenance-free:, innovative design lets you use this cell for determination of water in oils.
  • Cell with a diaphragm is recommended for applications such as the determination of water in substances containing ketones, also recommended for best possible accuracy
Coulometric Reagent Exchange Set
Allows emptying and re-filling the coulometric cell, reducing the risk of the user coming into contact with reagents

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Type Coulometric Cell without Diaphragm
For Use With (Application) General coulometric KF titration
Material Glass
For Use With (Equipment) C20 Coulometric Titrator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.