Learn More
Drummond™ Replacement Parts Kits for Drummond Microdispensers
GSA_VA
Catalog No. 211716
Change view
Click to view available options
Volume (Metric):
10 to 50μL
100 to 1000μL
2 to 10μL
20 to 100μL
5 to 25μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-171-6 2 to 10μL
21-171-7 5 to 25μL
21-171-9 20 to 100μL
21-171-8 10 to 50μL
22-231647 100 to 1000μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 21-171-6 Supplier Drummond™ Supplier No. 3000010K
Add to Cart
Add to Cart
Replacement Parts Kits. For Drummond Digital and Dialamatic Microdispensers.
Order Info
For contents of each kit, see chart.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Type Replacement Parts Kit
Volume (Metric) 2 to 10μL
For Use With (Equipment) Drummond Microdispenser
Includes 2 Plungers, 2 O-rings, 1 collet, 1 coil spring, 1 spring guide