Learn More
MilliporeSigma™ Replacement Parts for Stirred Cells
Catalog No. 8200SCPKIT Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With (Equipment):
Amicon 8003 Series Stirred Cell
Amicon 8200 Series Stirred Cell
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
8200SCPKIT Amicon 8200 Series Stirred Cell
8003SCPKIT Amicon 8003 Series Stirred Cell
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8200SCPKIT Supplier MilliporeSigma™ Supplier No. 8200SCPKIT
Suitable for use with Amicon™ stirred cell apparatus
  • Small Replacement Parts for 8000 Series Stirred Cells include 2 O-rings (Mfr. Nos. 2122S, 2118S, 2015S), tubing (Mfr. No. 4500202), tubing fitting assembly (Mfr. No. 15025AM, and membrane holder (Mfr. No. 2214801)
  • Large Replacement Parts for Stirred Cells include one each: cap assembly, stirrer assembly and base
TRUSTED_SUSTAINABILITY
Specifications
Includes Cap assembly (1524901), stirrer assembly (15247) and base (210000)
Type Replacement Part
For Use With (Equipment) Amicon 8200 Series Stirred Cell