Learn More
World Precision Instrument Replacement Pipette Holder for 5430-ALL
SDP
Catalog No. 502521225
Encompass
This part number is made up of (6) of the clear plastic pipette holders that go on the end of the 5430-ALL PicoNozzle. It does not include washers (seals) or the little metal cap.
Catalog No. 50-252-1225 Supplier World Precision Instrument Supplier No. 751256
This part number is made up of (6) of the clear plastic pipette holders that go on the end of the 5430-ALL PicoNozzle. It does not include washers (seals) or the little metal cap.