Learn More
Perkin Elmer US LLC Replacement Plastic Plungers for Gauze Loading Rig, Pkg. 5
Catalog No. 502112054
Encompass_Preferred
Package of five replacement plungers for the Gauze Loading Device (L4070023), which correctly positions gauze at both ends of stainless steel sampling tubes and is highly recommended when packing sample tubes.
Catalog No. 50-211-2054 Supplier Perkin Elmer US LLC Supplier No. L4071151
Package of five replacement plungers for the Gauze Loading Device (L4070023), which correctly positions gauze at both ends of stainless steel sampling tubes and is highly recommended when packing sample tubes.