Learn More
World Precision Instrument Replacement Platinum Temperature Probe for Air-Therm
SDP
Catalog No. 502521487
Encompass
  • Precision, shielded probe
  • Use with Air-Therm ATX Heater Controller 
  • Not for use with the Air-Therm SMT
Catalog No. 50-252-1487 Supplier World Precision Instrument Supplier No. 300276
  • Precision, shielded probe
  • Use with Air-Therm ATX Heater Controller 
  • Not for use with the Air-Therm SMT