Learn More
World Precision Instrument Replacement Plunger 2.0
SDP
Catalog No. 502539302
Encompass
The Rapid-Punch biopsy punch consists of a razor sharp cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin substrates. This punch is reusable, and these replacement plungers are designed for use with the 504639 Rapid-Punch Biopsy Punch. Choose the replacement biopsy punch plunger based on the size of your biopsy punch tip. Sizes include  0.5, 1.0, 1.2, 2.0 or 3.0 mm diameter. These replacement plungers are sold individually.
Catalog No. 50-253-9302 Supplier World Precision Instrument Supplier No. 504738
The Rapid-Punch biopsy punch consists of a razor sharp cutting tip designed to cut, retrieve and store cored samples from source materials such as tissue, gels, paper, cloth, leaves, paint chips, films or other thin substrates. This punch is reusable, and these replacement plungers are designed for use with the 504639 Rapid-Punch Biopsy Punch. Choose the replacement biopsy punch plunger based on the size of your biopsy punch tip. Sizes include  0.5, 1.0, 1.2, 2.0 or 3.0 mm diameter. These replacement plungers are sold individually.