Learn More
Analytical Sales & Services Replacement Pod for VXH593-888 - Rheodyne SS 2 Position, 6 Port Ultra High Pressure Switching Valve
Catalog No. 5000805784
Encompass
Replacement Pod for VXH593-888 - Rheodyne Stainless Steel 2 Position, 6 Port Ultra High Pressure Switching Valve. 15,000psi (1034 bar)\n\nQty/Pkg: EACH
Catalog No. 50-008-05784 Supplier Analytical Sales & Services Supplier No. PD8888
Replacement Pod for VXH593-888 - Rheodyne Stainless Steel 2 Position, 6 Port Ultra High Pressure Switching Valve. 15,000psi (1034 bar)\n\nQty/Pkg: EACH