Learn More
Analytical Sales & Services Replacement Pod for VXV7788-000 - PEEK 2 Position, 6 Port High Pressure Switching Valve
Catalog No. 5000805783
Encompass
Replacement Pod for VXV7788-000 - PEEK 2 Position, 6 Port High Pressure Switching Valve. 5,000psi (345 bar)\n\nQty/Pkg: EACH
Catalog No. 50-008-05783 Supplier Analytical Sales & Services Supplier No. PD7788
Replacement Pod for VXV7788-000 - PEEK 2 Position, 6 Port High Pressure Switching Valve. 5,000psi (345 bar)\n\nQty/Pkg: EACH