Learn More
MSC Extract-All Air Cleaner Filter, Polyester, 20 in. x 20 in., MERV 6, 6/Pk.
Catalog No. 5000508235
Encompass
These polyester panel filters are designed as replacements for clean air stations. Each pack contains 6 units constructed with durable polyester media to capture common airborne particles.
  • Features a MERV 6 performance rating for consistent filtration.
  • Designed to filter out dust, smoke, pollen, mites, and fibers.
  • Includes an integrated frame for ease of installation.
  • Tested in accordance with the ASHRAE 52 test method.
  • Constructed with a media area of 2.8 sq ft per filter.
Catalog No. 50-005-08235 Supplier MSC Supplier No. 60812732
These polyester panel filters are designed as replacements for clean air stations. Each pack contains 6 units constructed with durable polyester media to capture common airborne particles.
  • Features a MERV 6 performance rating for consistent filtration.
  • Designed to filter out dust, smoke, pollen, mites, and fibers.
  • Includes an integrated frame for ease of installation.
  • Tested in accordance with the ASHRAE 52 test method.
  • Constructed with a media area of 2.8 sq ft per filter.