Learn More
Electron Microscopy Sciences Replacement Needle for Probe #4
SDP
Catalog No. 50996783
Encompass
Replacement probe tip for needle probe. Made of hardened and nickel plated tool steel with a sharp tip.
Catalog No. 50-996-783 Supplier Electron Microscopy Sciences Supplier No. 6213004
Replacement probe tip for needle probe. Made of hardened and nickel plated tool steel with a sharp tip.