Learn More
SKC Inc PUF Replacement, 22-mm diameter x 76-mm length, uncleaned, for Low-volume PUF Tube, 20/pack
SDP
Catalog No. NC1542197
Encompass
SKC precleaned and ready-to-use Low-volume PUF Tubes contain polyurethane foam (PUF) or PUF/sorbent combinations that meet the specifications of EPA and ASTM indoor and ambient air sampling methods for pesticides in homes, public buildings, and offices. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. NC1542197 Supplier SKC Inc Supplier No. P22692
SKC precleaned and ready-to-use Low-volume PUF Tubes contain polyurethane foam (PUF) or PUF/sorbent combinations that meet the specifications of EPA and ASTM indoor and ambient air sampling methods for pesticides in homes, public buildings, and offices. DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”