Learn More
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Specifications
| Color | White |
| Composition | Boric Acid 45 to 50%, Calcium Thioglycollate Trihydrate 20 to 25%, Nonhazardous components 10 to 20%, Disodium Tetraborate 5 to 10% |
| pH Range | 8.4 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | Colorimeter and spectrophotometer |
| Quantity | 100/Pk. |
| Range | 1 to 10mg/L |
| Type | Iron HR Tablet |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.