Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+

Catalog No. p-8665000 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Iron HR Tablet
Silica No. 1 Tablet
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
09-053-190 Iron HR Tablet
09-053-194 Silica No. 1 Tablet
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-053-190 Supplier Lovibond™ Supplier No. 515380BT
Add to Cart
Add to Cart

Lovibond™ Replacement Reagent for CHECKIT™ Comparator 2000+ is manufactured under tightly controlled conditions to maintain the highest quality standards.

  • Highest quality production standards
  • Consistent and reliable reagents are provided in liquid and tablet formats
  • Maximum accuracy

Specifications

Color White
Composition Boric Acid 45 to 50%, Calcium Thioglycollate Trihydrate 20 to 25%, Nonhazardous components 10 to 20%, Disodium Tetraborate 5 to 10%
pH Range 8.4
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Quantity 100/Pk.
Range 1 to 10mg/L
Type Iron HR Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.