Learn More
IBI SCIENTIFIC 3P Replacement Rocker Cam, Facilitates the adjustment of rocking angle.
Catalog No. NC3665072
Encompass_Preferred
Located on outside of power moduleFacilitates the adjustment of rocking angle.Attaches directly to drive shaftThe ROCPL0002 contains one replacement drive cam for the Hi-Lo Rocker. This cam is located on the outside of the power module and facilitates the adjustment of the degree of angle for the Hi-Lo Rocker. The can connects to the drive shaft protruding from the power module.Made in the USA
Catalog No. NC3665072 Supplier IBI SCIENTIFIC 3P Supplier No. ROCPL0002
Located on outside of power moduleFacilitates the adjustment of rocking angle.Attaches directly to drive shaftThe ROCPL0002 contains one replacement drive cam for the Hi-Lo Rocker. This cam is located on the outside of the power module and facilitates the adjustment of the degree of angle for the Hi-Lo Rocker. The can connects to the drive shaft protruding from the power module.Made in the USA