Learn More
Perkin Elmer US LLC Replacement Rotor for 1/8 6-Port Valve, Temperature T
Catalog No. 502112852
Encompass_Preferred
The T type rotor material is designed for use in high-temperature applications, requiring the valve to be at or above 300°C. In order to seal properly and turn easily, after installation, the rotor should be temperature conditioned by brining the valve assembly to 325°C and cycling the valve (on-off) 10 times. The temperature of the valve then can be set according to the requirements of the user.
Catalog No. 50-211-2852 Supplier Perkin Elmer US LLC Supplier No. N9302905
The T type rotor material is designed for use in high-temperature applications, requiring the valve to be at or above 300°C. In order to seal properly and turn easily, after installation, the rotor should be temperature conditioned by brining the valve assembly to 325°C and cycling the valve (on-off) 10 times. The temperature of the valve then can be set according to the requirements of the user.