Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Replacement Rubber Adapter
GREENER_CHOICESDP

Catalog No. 31502295 Shop All Kemtech Products
Change view
Click to view available options
Material:
Rubber
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
31-502-295 Rubber
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 31-502-295 Supplier Synthware™ Supplier No. R045
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with gas inlet tubes or plain stem thermometers.

  • Replacement rubber adapter
  • For use with gas inlet tubes or plain stem thermometers

Specifications

Color Black
For Use With (Application) Connecting Two Pieces of Glassware
For Use With (Equipment) Gas Inlet Tubes, Plain Stem Thermometers
Material Rubber
Product Type Adapter
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.