Learn More
Grainger Replacement Saw Blade 16 In
Catalog No. 502766316
Encompass_Preferred
The Tri-Cut Saw Blade is a 16 inch circular saw blade made by Tri-Cut. It features a 5/8 inch arbor hole and is designed for cutting wood, plywood, and chipboard. The blade has 80 carbide tipped teeth which provide a smooth cut. It is .095 inches thick and can cut at speeds up to 5,900 RPM. The blade is manufactured in the USA.
Catalog No. 50-276-6316 Supplier Grainger Supplier No. 16X105
The Tri-Cut Saw Blade is a 16 inch circular saw blade made by Tri-Cut. It features a 5/8 inch arbor hole and is designed for cutting wood, plywood, and chipboard. The blade has 80 carbide tipped teeth which provide a smooth cut. It is .095 inches thick and can cut at speeds up to 5,900 RPM. The blade is manufactured in the USA.