Learn More
Analytical Sales & Services Replacement Screws for Nano Nest, 1-1/8"
Catalog No. 5000806723
Encompass
Replacement 18-8 Stainless Steel Socket Head Screw, 8-32 Thread Size, 1-1/8" Long\n\nQty/Pkg: 100
Catalog No. 50-008-06723 Supplier Analytical Sales & Services Supplier No. 118NNSCREW
Replacement 18-8 Stainless Steel Socket Head Screw, 8-32 Thread Size, 1-1/8" Long\n\nQty/Pkg: 100